Learn More
anti-ErbB3/Her3, PE, Clone: 66223, R&D Systems™
Manufacturer:  R&D Systems FAB3481P100
 View more versions of this product
Catalog No. FAB3481P100
Add to cart
Specifications
Specifications
ErbB3/Her3
66223
Flow Cytometry 10 uL/10^6 cells
P21860
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human ErbB3/Her3 in direct ELISAs. Does not cross-react with recombinant human EGF R.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
ERBB3
Mouse myeloma cell line NS0-derived recombinant human ErbB3/Her3, Ser20-Thr643, Accession # P21860
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2065
Human