Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB4, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120922

Catalog No. PA120922

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ErbB 4 is the fourth member of class I receptor kinase family. EbbB 4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDAMB-330 and MDA-MB-361. Expression of ErbB 4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3.

PA1-20922 detects ErbB4 in Human and Mouse samples.PA1-20922 has been successfully used in Immunohistochemistry (Paraffin), Immunoprecipitation and Western Blot procedures.PA1-20922 immunogen corresponds to Synthetic peptide corresponding to the C-terminal region of the human protein..
Specifications

Specifications

ErbB4
Polyclonal
Unconjugated
ERBB4
Purified
ERBB4
Synthetic peptide corresponding to the C-terminal region of the human protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
Q15303, Q61527
HER4, MGC138404, p180erbB4
Rabbit
IgG
500μL
Store at 4°C, do not freeze
Primary
2066
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit