Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
excision repair cross-complementing rodent repair deficiency, complementation group 3 (xeroderma pigmentosum group B complementing), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ERCC3.
Supplier: Abnova Corporation H00002071A01
Description
ERCC3 is an ATP-dependent DNA helicase that functions in nucleotide excision repair and complements xeroderma pigmentosum group B mutations. It also is the 89kDa subunit of basal transcription factor 2 (TFIIH) and thus functions in class II transcription. [provided by RefSeq
Sequence: APGNDPQEAVPSAAGKQVDESGTKVDEYGAKDYRLQMPLKDDHTSRPLWVAPDGHIFLEAFSPVYKYAQDFLVAIAEPVCRPTHVHEYKLTAYSLYAAVSSpecifications
excision repair cross-complementing rodent repair deficiency, complementation group 3 (xeroderma pigmentosum group B complementing) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant ERCC3. | |
ERCC3 | |
BTF2/GTF2H/RAD25/TFIIH/XPB | |
Mouse | |
50 μL | |
Yes | |
2071 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC008820 | |
ERCC3 | |
ERCC3 (AAH08820, 29 a.a. ∼ 128 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction