Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ERCC8, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132166

Catalog No. NBP132166


Add to cart

Description

Description

ERCC8 Polyclonal antibody specifically detects ERCC8 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Functional.
Specifications

Specifications

ERCC8
0.94mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Functional
Affinity Purified
ERCC8
Recombinant protein encompassing a sequence within the center region of human ERCC8. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Not yet tested in any other species.
Functional Assay, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8
Rabbit
IgG
0.1 ml
Cancer, DNA Repair, Nucleotide Excision Repair
Primary
1161
Human, Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only