Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524436

Catalog No. PIPA524436

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a WD repeat protein, which interacts with Cockayne syndrome type B (CSB) protein and with p44 protein, a subunit of the RNA polymerase II transcription factor IIH. Mutations in this gene have been identified in patients with hereditary disease Cockayne syndrome (CS). CS cells are abnormally sensitive to ultraviolet radiation and are defective in the repair of transcriptionally active genes.
Specifications

Specifications

ERCC8
Polyclonal
PBS with 0.09% sodium azide
Q13216, Q8CFD5
CKN1, CSA, Cockayne syndrome WD-repeat protein CSA, DNA excision repair protein ERCC-8, cockayne syndrome WD repeat protein CSA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1161, 71991
Immunohistochemistry, Western Blot
Unconjugated
ERCC8
Liquid
ERCC8
KLH conjugated synthetic peptide between 210-238 amino acids from the central region of human ERCC8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit