Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSPT2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526667

Catalog No. PIPA526667

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ERF3B encodes a GTPase that belongs to the GTP-binding elongation factor family. The encoded protein is a polypeptide release factor that complexes with eukaryotic peptide chain release factor 1 to mediate translation termination. This protein may also be involved in mRNA stability.
Specifications

Specifications

GSPT2
Polyclonal
PBS with 0.09% sodium azide
Q8IYD1, Q149F3
ERF3B, GST2, eukaryotic peptide chain release factor GTP-binding subunit ERF3B, eukaryotic peptide chain release factor subunit 3b
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14853, 23708
Western Blot
Unconjugated
GSPT2
Liquid
GSPT2
KLH conjugated synthetic peptide between 216-245 amino acids from the central region of human ERF3B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit