Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERG Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124327

Catalog No. PIPA124327


Add to cart

Description

Description

PA1-24327 detects ERG in human samples. PA1-24327 has been successfully used in Western blot procedures. The calculated molecular weight = 54 kDa. Differences in calculated versus apparent molecular weight may be due to post-translational modifications or protein hydrophobicity. The PA1-24327 immunogen is a synthetic peptide corresponding to residues 228-277 of human ERG.

v-ets erythroblastosis virus E26 oncogene like isoform 2 (ERG) is a transcriptional regulator. It may participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.
Specifications

Specifications

ERG
Polyclonal
Unconjugated
ERG
Liquid
ERG
Synthetic peptide corresponding to residues 228-277 of human ERG.
Protein A/G
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.2
P11308
erg-3, p55
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2078
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit