Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERGIC3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526226

Catalog No. PIPA526226

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse, non-human primate and zebrafish based on sequence homology.

ERGIC3 is possible role in transport between endoplasmic reticulum and Golgi.
Specifications

Specifications

ERGIC3
Polyclonal
PBS with 0.09% sodium azide
Q9Y282
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2, endoplasmic reticulum-Golgi intermediate compartment protein 3, endoplasmic reticulum-localized protein ERp43, serologically defined breast cancer antigen 84, serologically defined breast cancer antigen NY-BR-84
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51614
Immunohistochemistry, Western Blot
Unconjugated
ERGIC3
Liquid
ERGIC3
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ERGIC3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit