Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK3 Mouse, Unlabeled, Clone: 30, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610105

Catalog No. BDB610105


Add to cart

Description

Description

The family of serine/threonine kinases known as ERKs (extracellular signal regulated kinases) or MAPKs (mitogen-activated protein kinases) are activated after cell stimulation by a variety of hormones and growth factors. Cell stimulation induces a signaling cascade that leads to phosphorylation of MEK (MAPK/ERK kinase) which, in turn, activates ERK via tyrosine and threonine phosphorylation. A myriad of proteins represent the downstream effectors for the active ERK and implicate it in the control of cell proliferation and differentiation, as well as regulation of the cytoskeleton. Activation of ERK is normally transient and cells possess dual specificity phosphatases that are responsible for its down-regulation. Furthermore, multiple studies have shown that elevated ERK activity is associated with some cancers. ERK3 is a 63kDa family member that is 43% homologous to ERK1 and ERK2. A 97kDa ERK3 isoform has also been described which possesses an additional 178 amino acids.

Host Species: Mouse
Clone: 30
Isotype: IgM
Species Reactivity: Mouse
Immunogen: Rat ERK3 aa. 399-543
Formula Weight [Chemical]: 62kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

ERK3
30
Unconjugated
Murine
IgM
RUO
Primary
Canine, Chicken, Human, Murine, Rat
Immunofluorescence, Immunoprecipitation, Western Blot
250μg/mL
Affinity Purified
Rat ERK3 aa. 399-543
50μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit