Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERMN Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524818

Catalog No. PIPA524818


Add to cart

Description

Description

ERMIN plays a role in cytoskeletal rearrangements during the late wrapping and/or compaction phases of myelinogenesis as well as in maintece and stability of myelin sheath in the adult. ERMIN may play an important role in late-stage oligodendroglia maturation, myelin/Ranvier node formation during CNS development, and in the maintece and plasticity of related structures in the mature CNS.
Specifications

Specifications

ERMN
Polyclonal
PBS with 0.09% sodium azide
Q8TAM6
JN, KIAA1189, ermin, ermin, juxtanodin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57471
Flow Cytometry, Western Blot
Unconjugated
ERMN
Liquid
ERMN
KLH conjugated synthetic peptide between 113-141 amino acids from the central region of human ERMIN
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit