Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERO1L Rabbit anti-Human, Mouse, Rat, DyLight® 488, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522744

Catalog No. PIPA522744

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This ERO1L antibody is useful for Western Blot, where a band ~57 kDa is observed. Depending on the samples used ERO1L can also appear as a doublet which is believed to represent the oxidized and reduced forms of the protein. Suggested positive control: 293T, INS-1, and MIN6 cell lysate.

Perhaps the most distinctive feature of protein folding in the ER is the abundance of disulfide bonds that must form during maturation of proteins traveling along the secretory pathway. Formation of disulfide bonds is a redox reaction. Thus, to match the flux of disulfide bonds that exit from the ER by virtue of protein secretion, a flux of oxidizing equivalents into the ER is required. In eukaryotic cells, the essential protein relay supporting this flux, and hence disulfide bond formation, involves endoplasmic reticulum oxidoreductin 1 (Ero1) and protein disulfide isomerase (PDI). The temporal pattern of hypoxic ERO1-L alpha induction is very similar to that of genes triggered by the hypoxia inducible transcription factor (HIF-1) and is characteristically mimicked by cobalt and by deferoxamine, but is absent in cells with a defective aryl hydrocarbon receptor translocator (ARNT, HIF-1 alpha). We speculate from these findings that the expression of ERO1-L alpha is probably regulated via the HIF-pathway and thus belongs to the family of classic oxygen regulated genes.
Specifications

Specifications

ERO1L
Polyclonal
DyLight™ 488
ERO1A
Liquid
ERO1A, Ero1l
A synthetic peptide made the the N-terminal region of the human protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
2 mg/mL
sodium borate with 1% BSA and no preservative
Q96HE7, Q8R180, Q8R4A1
UNQ434/PRO865, ERO1-alpha, ERO1A, ERO1-L, ERO1-L-alpha, ERO1-like protein alpha, endoplasmic oxidoreductin-1-like protein, oxidoreductin-1-L-alpha
Rabbit
IgG
100 μL
4° C, store in dark
Primary
171562, 30001, 50527
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit