Learn More
ERO1L Rabbit anti-Human, Mouse, Rat, DyLight® 650, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522743
Catalog No. PIPA522743
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This ERO1L antibody is useful for Western Blot, where a band ~57 kDa is observed. Depending on the samples used ERO1L can also appear as a doublet which is believed to represent the oxidized and reduced forms of the protein. Suggested positive cont
Perhaps the most distinctive feature of protein folding in the ER is the abundance of disulfide bonds that must form during maturation of proteins traveling along the secretory pathway. Formation of disulfide bonds is a redox reaction. Thus, to match the flux of disulfide bonds that exit from the ER by virtue of protein secretion, a flux of oxidizing equivalents into the ER is required. In eukaryotic cells, the essential protein relay supporting this flux, and hence disulfide bond formation, involves endoplasmic reticulum oxidoreductin 1 (Ero1) and protein disulfide isomerase (PDI). The temporal pattern of hypoxic ERO1-L alpha induction is very similar to that of genes triggered by the hypoxia inducible transcription factor (HIF-1) and is characteristically mimicked by cobalt and by deferoxamine, but is absent in cells with a defective aryl hydrocarbon receptor translocator (ARNT, HIF-1 alpha). We speculate from these findings that the expression of ERO1-L alpha is probably regulated via the HIF-pathway and thus belongs to the family of classic oxygen regulated genes.
Specifications
Specifications
ERO1L
Polyclonal
DyLight™ 650
ERO1A
Liquid
Ero1a, Ero1l
A synthetic peptide made the the N-terminal region of the human protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
2 mg/mL
sodium borate with 1% BSA and no preservative
Q96HE7, Q8R180, Q8R4A1
UNQ434/PRO865, ERO1-alpha, ERO1A, ERO1-L, ERO1-L-alpha, ERO1-like protein alpha, endoplasmic oxidoreductin-1-like protein, oxidoreductin-1-L-alpha
Rabbit
IgG
100 μL
4° C, store in dark
Primary
171562, 30001, 50527