Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERp72 Mouse, Unlabeled, Clone: 3, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610970

Catalog No. BDB610970


Add to cart

Description

Description

Within the ER lumen, numerous molecular chaperones transiently bind nascent proteins and facilitate their folding or assembly. The action of chaperones ensures proper folding and, in turn, transport of new proteins from the rough ER to the Golgi complex. Some chaperones (grp78/Bip) bind to exposed hydrophobic segments, while others (ERp59/PDI) are involved in disulfide isomerization. An ERp59/PDI related protein, ERp72, contains three copies of the PDI acitve site, CGHC. Although ERp72 participates in disulfide isomerization, it is unclear whether it functions alone or in conjunction with other chaperones, specifically other members of the CGHC-containing protein family. Many chaperones, including PDI, are held within the ER via the common C-terminalkDaEL sequence. ERp72 contains a related KEEL sequence that is essential for its ER retention. Additional studies have shown that ERp72 is a protease and a member of the thioredoxin superfamily known to bind Ca2+;. Thus, ERp72 functions as a Ca2+;-dependent chaperone with disulfide isomerization activity, but also may undertake other, yet undefined, roles within the ER.

Immunofluorescence, Western Blotting

Specifications

Specifications

ERp72
3
Unconjugated
Affinity Purified
Human ERp72 aa. 427-642
50μg
Cell Biology
Primary
Human, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit