Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRRG Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527755

Catalog No. PIPA527755


Add to cart

Description

Description

PA5-27755 targets ERR gamma in WB applications and shows reactivity with Human and mouse samples. The PA5-27755 immunogen is a recombit fragment corresponding to a region within amino acids 53 and 435 of ERR gamma. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Orphan receptor that acts as transcription activator in the absence of bound ligand. Binds specifically to an estrogen response element and activates reporter genes controlled by estrogen response elements.
Specifications

Specifications

ESRRG
Polyclonal
Unconjugated
ESRRG
Liquid
ESRRG
Recombit fragment contain a sequence corresponding to a region within amino acids 53 and 435 of ERR gamma
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
0.51 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P62508, P62509, P62510
ERR3, ERRgamma, NR3B3, ERR gamma-2, estrogen receptor-related protein 3, nuclear receptor subfamily 3 group B member 3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2104, 26381, 360896
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit