Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Erythropoietin R, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $361.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide made to an internal portion of the human EPO receptor protein (between residues 300-400) [UniProt P19235]
Antigen Erythropoietin R
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP119388SS
View Documents
Novus Biologicals
NBP119388SS
0.025 ml
Each for $119.60
Add to cart
 
NBP119388
View Documents
Novus Biologicals
NBP119388
0.1 ml
Each for $361.50
Add to cart
 
Description

Description

Erythropoietin R Polyclonal antibody specifically detects Erythropoietin R in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
A synthetic peptide made to an internal portion of the human EPO receptor protein (between residues 300-400) [UniProt P19235]
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
P19235
PBS with 0.02% Sodium Azide
IgG
Erythropoietin R
Affinity Purified
RUO
EpoR (A364) detects endogenous levels of EpoR protein.
2057
1.0 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit