Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EpoR, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Supplier:  Thermo Scientific PA120180

Catalog No. PIPA120180

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Recombinant Human Erythropoietin Receptor (EPO R) is a type I membrane-spanning protein, expressed in a mouse myeloma cell line, NSO. This product is produced from a cDNA sequence encoding the extracellular domain of the EPO receptor. The 225 amino acid residue mature recombinant soluble EPO receptor, generated after cleavage of a 25 amino acid residue signal peptide, has predicted molecular mass of approximately 25 kDa.

PA1-20180 detects EPO Receptor in human samples.PA1-20180 has been successfully used in ELISA and Western blot procedures. Western blot detects a band of ∽50 kDa.The PA1-20180 immunogen is recombinant, full length, human EPO receptor, expressed in mouse myeloma NSO cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Erythropoietin Receptor
Polyclonal
Unconjugated
P19235
EPOR
Recombinant, full length, human EPO receptor, expressed in mouse myeloma NSO cells.
25 μg
Primary
Human
Antibody
IgG
ELISA, Western Blot
0.1mg/mL
EPOR
MGC138358
Goat
Antigen affinity chromatography
RUO
2057
Store at -20°C or Store at -80°C if preferred
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit