Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESAM Rat anti-Mouse, Clone: 340236, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524072

Catalog No. PIMA524072


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

ESAM
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q925F2
2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
69524
Immunohistochemistry (Frozen)
340236
Unconjugated
Esam
Lyophilized
ESAM
Mouse myeloma cell line NS0-derived recombit mouse ESAM Gln30-Ser248 (predicted)
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit