Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESCO2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524210

Catalog No. PIPA524210


Add to cart

Description

Description

This gene encodes a protein that may have acetyltransferase activity and may be required for the establishment of sister chromatid cohesion during the S phase of mitosis. Mutations in this gene have been associated with Roberts syndrome.
Specifications

Specifications

ESCO2
Polyclonal
Unconjugated
ESCO2
Liquid
ESCO2
KLH conjugated synthetic peptide between 132-161 amino acids from the central region of human ESCO2
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
0.25 mg/mL
PBS with 0.09% sodium azide
Q56NI9, Q8CIB9
2410004I17Rik, EFO2, RBS, ECO1 homolog 2, N-acetyltransferase ESCO2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
157570, 71988
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit