Learn More
anti-ESD, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232344
Catalog No. NBP232344
Add to cart
Description
Description
ESD Polyclonal antibody specifically detects ESD in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
ESD
0.5 mg/mL
Western Blot 1:1000
P10768
EC 3.1.2.12, esterase 10, esterase Desterase D/formylglutathione hydrolase, FGH, FLJ11763, S-formylglutathione hydrolase
Rabbit
IgG
Protein A purified
RUO
Primary
2098
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
ESD
This ESD antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 68-102 amino acids from the Central region of human ESD.
31.463 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat