Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525833

Catalog No. PIPA525833


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

RBM35A, also known as ESRP1, is a mRNA splicing factor that with its related protein RBM35B (ESRP2) are coordinators of an epithelial cell-type-specific splicing program. RBM35A contains three putative RNA recognition motifs and acts by directly binding specific sequences in mRNAs. RBM35A is involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5 ft. UTRs of mRNAs. Other recent studies have shown that RMB35A may also act as a novel tumor suppressor.
Specifications

Specifications

ESRP1
Polyclonal
PBS with 0.09% sodium azide
Q6NXG1
RBM35A, RMB35A, RNA binding motif protein 35A, RNA-binding motif protein 35A, RNA-binding protein 35A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54845
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ESRP1
Liquid
ESRP1
KLH conjugated synthetic peptide between 182-211 amino acids from the N-terminal region of human ESRP1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit