Learn More
Estradiol Sheep anti-Chemical, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA175002
Catalog No. PIPA175002
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Estradiol (17ß-estradiol) is a sex hormone. Estradiol is the predomit sex hormone present in females. It is also present in males, being produced as an active metabolic product of testosterone. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.
Specifications
Specifications
Estradiol
Polyclonal
Unconjugated
Liquid
Sheep
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical
ELISA
8.61 mg/mL
PBS with 0.09% sodium azide; pH 7.2
17-Beta-Estradiol; 17-beta-hydroxysteroid dehydrogenase; 17-beta-HSD; Estrogen 17-oxidoreductase
17a-Estradiol(3)-BTG
Purified
RUO
Antibody
Polyclonal