Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen Receptor alpha Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137651

Catalog No. 01-673-425


Add to cart

Description

Description

This antibody is predicted to react with porcine based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is breast carcinoma.

Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis.
Specifications

Specifications

Estrogen Receptor alpha
Polyclonal
TBS with 1% BSA and 0.1% sodium azide; pH 7.6
P03372
ER, ESR, ESRA, Era, NR3A1, ER-alpha, estradiol receptor, estrogen nuclear receptor alpha, estrogen receptor, estrogen receptor alpha 3*,4,5,6,7*/822 isoform, estrogen receptor alpha delta 3*,4,5,6,7*,8*/941 isoform, estrogen receptor alpha delta 3*,4,5,6,7*/819-2 isoform, estrogen recepto
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2099
Immunohistochemistry (Paraffin)
Unconjugated
ESR1
Liquid
ESR1
Synthetic peptide derived from C-terminus of human estrogen receptor.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit