Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRRG Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1316

Catalog No. PA1316

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Estrogen-related receptor gamma (ERR gamma, ERR3, NR3B3) is an orphan member of the nuclear hormone receptor superfamily. All the ERR family members (1,2, and 3) share an almost identical DNA-binding domain, which has 68% amino acid identity with that of estrogen receptor. Studies involving the crystal structure of the LBD have shown a complex between ERR3 and SRC-1. ERR3 binds as a homodimer to direct repeats (5’-AGG TCA-3’), to extended half-sites, and to the inverted estrogen response element.The corresponding gene for the ERR3 is NR3B3.

PA1-316 detects human and mouse ERR3 from transfected cells and shows no cross-reactivity to ERR1.PA1-316 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~54kDa protein representing ERR1 from COS-7 transfected cells. This antibody is not suitable for 293T cells in Western blot analysis.PA1-316 immunogen is a synthetic peptide corresponding to residues D(401) Y E A G Q H M E D P R R(413) of human ERR3. This sequence is completely conserved between human, mouse and rat. This immunizing peptide (PEP-232) can be used for neutralization and control experiments.
Specifications

Specifications

ESRRG
Polyclonal
Unconjugated
ESRRG
Liquid
ESRRG
Synthetic peptide corresponding to residues D(401) Y E A G Q H M E D P R R(413) of human ERR3.
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P62508, P62509
Estrogen-Related Receptor Gamma, ERR gamma, NR3B3, Estrogen receptor-related protein 3, ERR gamma-2
Rabbit
IgG
200 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2104, 26381
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit