Learn More
Estrone Rabbit anti-Chemical, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA185741
Catalog No. PIPA185741
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-85741 is specific for oestrone. Less than 0.01% cross-reactivity was observed with oestradiol 17B, oestriol, progesterone and testosterone. Rabbit anti estrone antibody recognizes the aromatized C18 steroid, estrone.
Estrone is an aromatized C18 steroid with a 3-hydroxyl group and a 17-ketone, a major mammalian estrogen. it is converted from androstenedione directly, or from testosterone via estradiol. In humans, it is produced primarily by the cyclic ovaries, placenta, and the adipose tissue of men and postmenopausal women. Therapeutic estrone is the synthetic form of a naturally occurring estrogen estrone. Estrone diffuses through the cell membrane and binds to and subsequently activates the nuclear estrogen receptor found in the reproductive tract, breast, pituitary, hypothalamus, liver, and bone. The activated hormone-receptor complex gets translocated to the nucleus where it binds to the estrogen response element on the dna and promotes the transcription of target genes involved in the functioning of the female reproductive system and secondary sex characteristics. Estrone increases the hepatic synthesis of sex hormone binding globulin (shbg), thyroid-binding globulin (tbg), and other serum proteins, as well as suppresses the release of follicle-stimulating hormone (fsh) and luteinizing hormone (lh) from the anterior pituitary. [National Center for Biotechnology Information. PubChem Compound Database; CID=5870]
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Estrone
Polyclonal
Folliculin; OESTRONE; C18H22O2
Synthetic oestrone conjugated to BSA.
RUO
4°C or -20°C if preferred
Liquid
Chemical
ELISA
Unconjugated
Rabbit
100 μL
Primary
Antibody
IgG
Product Suggestions
Product Suggestions