Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETEA Goat anti-Bovine, Canine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131176

Catalog No. PIPA131176


Add to cart

Description

Description

PA1-31176 detects ETEA from human, mouse, cow, dog, and rat samples. PA1-31176 has been successfully used in Western blot and ELISA applications. The PA1-31176 immunogen is a synthetic peptide: HTEVLFVQDLTDE, corresponding to amino acids 433-445 of Human ETEA. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.
Specifications

Specifications

ETEA
Polyclonal
Unconjugated
FAF2
Liquid
FAF2
synthetic peptide HTEVLFVQDLTDE, corresponding to amino acids 433-445 of Human ETEA.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Rat
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q2HJD0, Q96CS3, Q3TDN2, Q5BK32
ETEA, KIAA0887, UBXD8, UBXN3B
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23197, 291000, 489102, 76577, 767948
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit