Learn More
ETNK1 Mouse anti-Human, Clone: 3F11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522149
Catalog No. PIMA522149
Add to cart
Description
Description
Peptide Sequence: WDPQEVTLQL FTDGITNKLI GCYVGNTMED VVLVRIYGNK TELLVDRDEE VKSFRVLQAH GCAPQLYCTF NNGLCYEFIQ GEALDPKHVC NPAIFR
This gene encodes an ethanolamine kinase, which functions in the first committed step of the phosphatidylethanolamine synthesis pathway. This cytosolic enzyme is specific for ethanolamine and exhibits negligible kinase activity on choline. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
Specifications
ETNK1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9HBU6
EKI, EKI 1, EKI1, Nbla10396, putative protein product of Nbla10396
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
55500
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
3F11
Unconjugated
ETNK1
Liquid
ETNK1
ETNK1 (NP_061108, 133 a.a. ∽ 228 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human