Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETNK1 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513823

Catalog No. PIPA513823

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ethanolamine kinase 1 (EKI1), functions in the first committed step of the phosphatidylethanolamine synthesis pathway. This cytosolic enzyme is specific for ethanolamine and exhibits negligible kinase activity on choline.
Specifications

Specifications

ETNK1
Polyclonal
PBS with 0.09% sodium azide
Q9HBU6, Q9D4V0
Ethanolamine kinase; EKI; ETNK1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100765795, 55500, 75320
Western Blot
Unconjugated
ETNK1
Liquid
ETNK1
KLH conjugated synthetic peptide between 356-386 amino acids from the C-terminal region of human Ethanolamine Kinase (EKI1)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit