Learn More
Ethinylestradiol Sheep anti-Chemical, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA175139
Catalog No. PIPA175139
Add to cart
Description
Description
PA1-75139 specifically detects Ethinylestradiol. PA1-75139 has been successfully used in ELISA applications. The PA1-75139 immunogen is Ethinylestradiol(6)-BTG.
Ethinyl Estradiol is a semisynthetic alkylated estradiol with a 17-alpha-ethinyl substitution. It has high estrogenic potency when administered orally, and is often used as the estrogenic component in oral contraceptives. Ethinyl estradiol binds to the estrogen receptor complex and enters the nucleus, activating DNA transcription of genes involved in estrogenic cellular responses. This agent also inhibits 5-alpha reductase in epididymal tissue, which lowers testosterone levels and may delay progression of prostatic cancer. In addition to its antineoplastic effects, ethinyl estradiol protects against osteoporosis. In animal models, short-term therapy with this agent has been shown to provide long-term protection against breast cancer, mimicking the antitumor effects of pregcy. [National Center for Biotechnology Information. PubChem Compound Database; CID=5991]
Specifications
Specifications
Ethinylestradiol
Polyclonal
PBS with 0.09% sodium azide
Ethinyl estradiol; Ethynyl estradiol; Ethynylestradiol; Ethinyloestradiol; C20H24O2
Ethinylestradiol(6)-BTG
Purified
RUO
Antibody
Polyclonal
ELISA
Unconjugated
Liquid
Sheep
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical