Learn More
ETS2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526845
Catalog No. PIPA526845
Add to cart
Description
Description
This antibody is predicted to react with bovine and mouse based on sequence homology.
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.
Specifications
Specifications
ETS2
Polyclonal
Unconjugated
ETS2
Liquid
ETS2
KLH conjugated synthetic peptide between 146-173 amino acids from the central region of human ETS2
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P15036
ETS2IT1, oncogene ETS-2, protein C-ets-2, v-ets avian erythroblastosis virus E2 oncogene homolog 2, v-ets avian erythroblastosis virus E26 oncogene homolog 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2114