Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETV5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524226

Catalog No. PIPA524226


Add to cart

Description

Description

ETS variant 5,transcription factor of the ETS family,divergent member of the winged helix-turn-helix superfamily,expressed in brain,placenta and other tissues involved in development. Tissue specificity: Ubiquitous. ERM is expressed in Sertoli cells and spermatogonial stem cells (SSCs). Studies on knockout mice have shown that it is required for SSC self-renewal but not differentiation, thus affected mice exhibit progressive germ cell depletion.
Specifications

Specifications

ETV5
Polyclonal
PBS with 0.09% sodium azide
P41161
ERM, ETS translocation variant 5, ets-related molecule, ets-related protein ERM
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2119
Flow Cytometry, Western Blot
Unconjugated
ETV5
Liquid
ETV5
KLH conjugated synthetic peptide between 8-36 amino acids from the N-terminal region of human ETV5
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit