Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
eIF2d Mouse anti-Human, Clone: 2D10, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518796
Catalog No. PIMA518796
Description
Description
Peptide Sequence: KGRICPIDIT LAQRASNKKV TVVRNLEAYG LDPYSVAAIL QQRCQASTTV NPAPGAKDSL QVQIQGNQVH HLGWLLLEEY QLPRKHIQGL EKALKPGKKK
This gene encodes a protein receptor that localizes phosphoglycoproteins within endosomes and at the cell periphery. This trafficking receptor for phosphoglycoproteins may play a role in neuroplasticity by modulating cell-cell interactions, intracellular adhesion, and protein binding at membrane surfaces. In hippocampal neurons, long-lasting down-regulation of ligatin mRNA levels occurs via post-transcriptional RNA processing following glutamate receptor activation. This protein contains single PUA and SUI1 domains and these domains may function in RNA binding and translation initiation, respectively.
Specifications
Specifications
eIF2d
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
P41214
Eukaryotic translation initiation factor 2D, HCA56, LGTN
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1939
ELISA, Western Blot
2D10
Unconjugated
EIF2D
Liquid
EIF2D
EIF2D (NP_008824.2, 485 a.a. ∽ 584 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human