Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVI5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526176

Catalog No. PIPA526176


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

EVI5 is a novel centrosomal protein with a complex expression pattern and subcellular localization, possibly involved in centrosome stability and dynamics. It may act as an oncogene.
Specifications

Specifications

EVI5
Polyclonal
PBS with 0.09% sodium azide
O60447, P97366
NB4S, EVI-5, dJ846D11.1 (ecotropic viral integration site 5), ecotropic viral integration site 5 protein homolog, neuroblastoma stage 4S gene protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
14020, 7813
Western Blot
Unconjugated
EVI5
Liquid
EVI5
KLH conjugated synthetic peptide between 191-220 amino acids from the central region of human EVI5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit