Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVX1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124495

Catalog No. PA124495


Add to cart

Description

Description

PA1-24495 detects EVX1 in Human samples. PA1-24495 has been successfully used in Immunohistochemistry and Western Blot procedures. PA1-24495 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human EVX1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

EVX1 is a member of the vertebrate eve-related homeo box family. This protein acts as a transcriptional repressor. A similar protein in mice is critical for early embryogenesis.
Specifications

Specifications

EVX1
Polyclonal
Unconjugated
EVX1
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2128
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P49640
EVX1
Synthetic peptide region of human EVX1.
Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit