Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EWSR1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127592

Catalog No. PA127592

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Might normally function as a repressor. EWS-fusion-proteins (EFPS) may play a role in the tumorigenic process. They may disturb gene expression by mimicking. or interfering with the normal function of CTD-POLII within the transcription initiation complex.

PA1-27592 detects EWSR1 from human, mouse, and rat samples.PA1-27592 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.Synthetic peptide corresponding to human RNA-binding protein EWSR1, amino acids 380-488
Specifications

Specifications

EWSR1
Polyclonal
PBS with no preservative
Q01844, Q61545
AC002059.7, EWS, bK984G1.4
Chicken
IgY
50μg
Store at -20°C
Primary
2130
Western Blot
Unconjugated
EWSR1
Purified
EWSR1
Synthetic peptide corresponding to human RNA-binding protein EWSR1, amino acids 380-488
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit