Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOC5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526024

Catalog No. PIPA526024

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

EXOC5 is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.
Specifications

Specifications

EXOC5
Polyclonal
PBS with 0.09% sodium azide
O00471
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P, SEC10-like 1, exocyst complex component Sec10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10640
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
EXOC5
Liquid
EXOC5
KLH conjugated synthetic peptide between 40-66 amino acids from the C-terminal region of human EXOC5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit