Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exportin-1 (CRM1) Mouse anti-Human, Unlabeled, Clone: 17, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611833

 View more versions of this product

Catalog No. BDB611833


Add to cart

Description

Description

The nuclear pore complex (NPC) is a 125MDa nuclear envelope complex that mediates nucleocytoplasmic shuttling of RNA and protein. NPC is comprised of more than 30 different proteins, including Ran/TC4, importins, and exportins . Exportin-1(hCRM1/XPO1) is the widely expressed human homologue of the yeast CRM1 protein, which functions in the maintenance of chromatin structure and nuclear export. Exportin-1 has an N-terminal CRIME domain that is homologous to yeast CRM1 and importin-β. Intracellular localization of Exportin-1 shows association with the NPC and overexpression of Exportin-1 in Xenopus oocytes stimulates Rev and U snRNA export from the nucleus. Exportin-1 also binds other nuclear transport related proteins, such as RanGTP, snurportin-1, and the nucleoporins CAN/Nup214 and Nup50. Overexpression of CAN leads to sequestration of Exportin-1 in the nucleoplasm, which inhibits cell growth and induces apoptosis. In addition, Exportin-1 may export specific proteins, such as Rev and PKI, via binding to a leucine-rich nuclear export signal (NES). Thus, Exportin-1 is an important protein and RNA nuclear export receptor, which may be required for normal cell growth and survival.

Host Species: Mouse
Clone: 17
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human hCRM1 aa. 2-122

Immunofluorescence, Western Blotting

Specifications

Specifications

Exportin-1 (CRM1)
17
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
CRM1
Affinity Purified
Human hCRM1 aa. 2-122
150μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.