Learn More
Exportin T Mouse anti-Human, Clone: LOS1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051525
Catalog No. 05-152-5MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-Exportin T Antibody, clone LOS1 (Mouse Monoclonal Antibody) validated in western blotting to detect Exportin T also known as tRNA (nuclear export receptor for tRNAs).
Specifications
Specifications
Exportin T
LOS1
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Purified
Murine
IgG1, κ, κ
100μg
Primary
Human
Western Blot
Unconjugated
NP_009166
XPOT, Exportin(tRNA), XPO3, exportin-T
Recombinant protein corresponding to Chlamydomonas Exportin-T.
Protein G purfied
2-8°C for one year
Monoclonal