Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EZH2/KMT6, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide made to an internal portion of the human KMT6/EZH2 protein (within residues 300-400). [Swiss-Prot# Q15910]
Antigen EZH2/KMT6
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11083976T
View Documents
Novus Biologicals
NB11083976SS
0.025 ml
Each for $119.60
Add to cart
 
NB11083976
View Documents
Novus Biologicals
NB11083976
0.1 ml
Each for $314.00
Add to cart
 
Description

Description

EZH2/KMT6 Polyclonal antibody specifically detects EZH2/KMT6 in Human, Mouse, Primate samples. It is validated for Western Blot.
Specifications

Specifications

Rabbit
Synthetic peptide made to an internal portion of the human KMT6/EZH2 protein (within residues 300-400). [Swiss-Prot# Q15910]
Polyclonal
Primary
Western Blot
2146
1 mg/mL
IgG
EZH2/KMT6
Affinity Purified
RUO
Q15910
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit