Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH2 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146476

Catalog No. PA146476


Add to cart

Description

Description

Suggested positive control: antigen standard for EZH2 (transient overexpression lysate), Jurkat whole cell lysate.

EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
Specifications

Specifications

EZH2
Polyclonal
Unconjugated
EZH2
Liquid
EZH2
Synthetic peptide corresponding to residues 300-400 of an internal portion of human EZH2. [Swiss-ProtNo. Q15910]
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate
Western Blot
1 mg/mL
tris citrate with 0.1% sodium azide; pH 7-8
Q15910, Q61188
Enhancer of zeste homolog 2 , EZH 2 , EZH-2 , ENX 1 , Enx-1 , Enx1h , an
Rabbit
IgG
100 μL
4° C, do not freeze
Primary
14056, 2146
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit