Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ezrin Mouse, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610602

 View more versions of this product

Catalog No. BDB610602


Add to cart

Description

Description

First described as an 80kDa protein concentrated in the apical cytoskeletal region of intestinal brush border cells, ezrin is now recognized as a major substrate of protein tyrosine kinases, such as the epidermal growth factor (EGF) tyrosine kinase. Ezrin is expressed at high levels in intestine, kidney, and placenta. In placenta, ezrin is present as monomers and non-covalent oligomers in tight association with actin microfilaments. In the human epidermoid carcinoma cell line A431, microvilli-like structures appear within 30 seconds after the addition of EGF. These structures give way to membrane ruffles after 2-5 minutes, followed by cell rounding after 10-20 minutes. At the same time, ezrin is recruited into these structures and oligomers are formed following its tyrosine phosphorylation. It is thought that tyrosine phosphorylation triggers the formation of ezrin oligomers.

Specifications

Specifications

Ezrin
18
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.