Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F-box protein 15/FBXO15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132860
Description
F-box protein 15/FBXO15 Polyclonal specifically detects F-box protein 15/FBXO15 in Human samples. It is validated for Western Blot.Specifications
F-box protein 15/FBXO15 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
FBXO15 | |
Recombinant protein encompassing a sequence within the center region of human FBXO15. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
201456 | |
Human | |
IgG |
Western Blot | |
1.0mg/mL | |
Western Blot 1:500-1:3000 | |
F-box only protein 15, F-box protein 15, FBX15MGC39671 | |
Rabbit | |
57 kDa | |
100 μL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction