Learn More
FBXO40 Mouse anti-Human, Clone: 2E5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA521977
Catalog No. PIMA521977
Add to cart
Description
Description
Peptide Sequence: QWKKKRYSHG GTSWRVHREI WQFSSLFSKI KSWEFNEVTS MSEHLKSCPF NIVEHKTDPI LLTSMCQPRE QARESLVSTF RIRPRGRYVS
Members of the F-box protein family, such as FBXO40, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).
Specifications
Specifications
FBXO40
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9UH90
burs-beta
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
51725
ELISA
2E5
Unconjugated
FBXO40
Liquid
FBXO40
FBXO40 (NP_057382, 620 a.a. ∽ 709 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human