Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM162A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524470
Description
Specifications
FAM162A | |
Polyclonal | |
FAM162A | |
C3orf28, E2IG5, HGTD-P, E2-induced gene 5 protein, HIF-1 alpha-responsive proapoptotic molecule, growth and transformation-dependent protein, protein FAM162A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26355, 70186 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q96A26, Q9D6U8 | |
FAM162A | |
KLH conjugated synthetic peptide between 118-146 amino acids from the C-terminal region of human F162A | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction