Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F1A alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130706

Catalog No. PIPA130706


Add to cart

Description

Description

PA1-30706 detects F1A alpha from mouse, human, rat samples. PA1-30706 has been successfully used in Western blot applications. The PA1-30706 immunogen is: Synthetic peptide: amino acids 609-622 of Human F1A alpha. This sequence is identical to the corresponding sequence of mouse. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

F1A alpha
Polyclonal
Unconjugated
FEM1B
Liquid
FEM1B
synthetic peptide amino acids 609-622 of Human F1A alpha. This sequence is identical to the corresponding sequence of mouse.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.02% sodium azide; pH 7.4
Q9UK73, Q9Z2G0, P0C6P7
DKFZp451E0710, FIAA
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10116, 14155, 315745
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit