Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80 Rat anti-Mouse, PE, Clone: BM8, Invitrogen™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MF48004

Catalog No. 50-113-8112


Add to cart

Description

Description

EGF-Like Module Containing Mucin-Like Hormone Receptor-Like Sequence 1 (EMR1) is characterized as an Orphan-B GPCR with an unknown ligand. The C-terminus contains seven hydrophobic segments, characteristic of a G protein-coupled receptor. The N-terminus of EMR1 has 6 EGF-like modules, separated from the transmembrane segments by a serine/threonine-rich domain, a feature reminiscent of mucin-like, single-span, integral membrane glycoproteins with adhesive properties. EMR1 has been reported to be expressed in human peripheral blood mononuclear cells, monocytes, and hematopoietic and solid tumor-derived cell lines. ESTs have been isolated from human normal spleen and mixed liver/spleen libraries and from human kidney, colon, brain, and bone marrow cancer libraries.

Specifications

Specifications

F4/80
Monoclonal
0.2mg/mL
PBS with 10% sucrose, 4mg/mL BSA and 0.1% sodium azide
Q61549
EMR1, EMR1 hormone receptor, F4/80, EGF-like module receptor 1, TM7LN3
Rat
IgG2a
500μL
4° C
Primary
13733
Flow Cytometry
BM8
PE
Adgre1
Liquid
Adgre1
mouse F4/80 which is a macrophage-restricted cell surface glycoprotein.
Purified
RUO
Antibody
Monoclonal
Murine
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit