Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM91A1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524948
Description
This antibody is predicted to react with bovine and chicken based on sequence homology.
Specifications
FAM91A1 | |
Polyclonal | |
FAM91A1 | |
protein FAM91A1, skeletal muscle cells re-entry induced | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
157769, 210998 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q658Y4, Q3UVG3 | |
FAM91A1 | |
KLH conjugated synthetic peptide between 127-155 amino acids from the N-terminal region of human F91A1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction