Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP1 Mouse anti-Human, Clone: 328607, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524055

Catalog No. PIMA524055


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human FABP2, -3, -4, -5, -6, -7, or recombit mouse FABP9 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
Specifications

Specifications

FABP1
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P07148
FABPL, L-FABP, fatty acid-binding protein 1, fatty acid-binding protein, liver, liver-type fatty acid-binding protein
Mouse
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2168
Immunohistochemistry (Paraffin), Western Blot
328607
Unconjugated
FABP1
Lyophilized
FABP1
E. coli-derived recombit human FABP1/L-FABP Met1-Ile127
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit