Learn More
fatty acid binding protein 1, liver, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FABP1 protein.
Supplier: Abnova Corporation H00002168B01
Description
FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. [provided by RefSeq
Sequence: MSFSGKYQLQSQENFEAFMKAIGLPEELIQKGKDIKGVSEIVQNGKHFKFTITAGSKVIQNEFTVGEECELETMTGEKVKTVVQLEGDNKLVTTFKNIKSVTELNGDIITNTMTLGDIVFKRISKRISpecifications
fatty acid binding protein 1, liver | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human FABP1 protein. | |
FABP1 | |
FABPL/L-FABP | |
Mouse | |
50 μL | |
Yes | |
2168 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC032801 | |
FABP1 | |
FABP1 (AAH32801, 1 a.a. ∼ 127 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.