Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524949

Catalog No. PIPA524949


Add to cart

Description

Description

This antibody is predicted to react with bovine and chicken based on sequence homology.

FABP7 is a brain fatty acid binding protein. Fatty acid binding proteins (FABPs) are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs are thought to play roles in fatty acid uptake, transport, and metabolism.
Specifications

Specifications

FABP7
Polyclonal
Unconjugated
FABP7
Liquid
FABP7
KLH conjugated synthetic peptide between 104-132 amino acids from the C-terminal region of human FABP7
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 0.09% sodium azide
O15540, P51880
B-FABP, BLBP, FABPB, MRG, brain lipid binding protein, brain lipid-binding protein, brain-type fatty acid-binding protein, fatty acid-binding protein 7, fatty acid-binding protein, brain, mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12140, 2173
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit