Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor IX Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522304

Catalog No. PIPA522304


Add to cart

Description

Description

PA5-22304 targets Factor IX in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-22304 immunogen is a recombit fragment corresponding to a region within amino acids 217 and 453 of Factor IX (Uniprot IDNo.P00740). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Factor IX
Polyclonal
Unconjugated
F9
Liquid
F9
Recombit fragment corresponding to a region within amino acids 217 and 453 of Factor IX (Uniprot IDNo.P00740)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Immunohistochemistry (Paraffin), Western Blot
0.95 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
P00740, P16296
RP6-88D7.1, FIX, HEMB, P19, PTC, Christmas factor, F9 p22, FIX F9, factor 9, factor IX F9, plasma thromboplastic component, plasma thromboplastin component, serine protease
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2158, 24946
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit